Learn More
Methanol with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend, Solstice
Shop All Solstice Research Chemicals ProductsSpecifications
Specifications
| Boiling Point | 64.7°C |
| CAS Max % | 99.8000%, 0.2000% |
| UN Number | UN1230 |
| Density | 0.792 g/cm3 |
| Grade | Solvent Blend |
| Packaging | Glass Bottle |
| Quantity | 4 x 4 L |
| Flash Point | 20°C |
| Chemical Name or Material | Methanol with 0.1% Trifluoroacetic acid, LabReady™ Solvent Blend |
Safety and Handling
| ShelfLife | 365 days from date of manufacture |
| DOTInformation | Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol Solution |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.