Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, 15% v/v Made with HPLC grade components, Reagents

Catalog No. 01334739
Change view
Click to view available options
:
Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
01-334-739 Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-739 Supplier Reagents Holdings Llc Supplier No. CM1207004C
Only null left
Add to Cart
Add to Cart

Methanol, 15% v/v Made with HPLC grade components, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 67-56-1
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name water
SMILES O

Specifications

Color Colorless
Boiling Point 98°C
Quantity 4 L
Packaging Glass Bottle
Physical Form Liquid
Specific Gravity 0.9705
Percent Purity 15%
Grade Reagent
Chemical Name or Material Methanol, 15% v/v Made with HPLC grade components
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.