Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, 99.8%, Extra Dry, anhydrous, SC, AcroSeal™, ACROS Organics™

Click to view available options
Quantity:
100mL
Packaging:
AcroSeal™ Glass Bottle in Tin Can
67-56-1
CH4O
32.042
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO
67-56-1
CH4O
32.042
MFCD00004595
OKKJLVBELUTLKV-UHFFFAOYSA-N
methyl alcohol,wood alcohol,carbinol,wood spirit,wood naphtha,methylol,methyl hydroxide,pyroxylic spirit,colonial spirit,columbian spirit
887
CHEBI:17790
methanol
CO
Additional Information Vapor Pressure: 128hPa at 20°C
Assay Percent Range 99.8%
Linear Formula CH3OH
Beilstein 01, IV, 1227
Fieser 11,322; 13,337
Merck Index 15, 6029
Formula Weight 32.04
Percent Purity ≥99.8%
Grade Extra Dry
Density 0.7910g/mL
Identification Pass Test
Packaging AcroSeal™ Glass Bottle in Tin Can
Water 50ppm max.
Quantity 100mL
Chemical Name or Material Methanol, Anhydrous
Show More Show Less

GHS H Statement:
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes damage to organs.
Highly flammable liquid and vapor.

GHS P Statement:
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Gently wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Danger