Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, ACS, absolute, low acetone, 99.8+%

Catalog No. AA31721M1
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA31721M1 4 x 1 L
AA31721AP 500 mL
AA31721K2 1 L
AA31721K7 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. AA31721M1 Supplier Thermo Scientific Chemicals Supplier No. 031721M1
Only null left
Add to Cart

CAS: 67-56-1 | CH4O | 32.04 g/mol

Methanol is useful in the synthesis of nucleic acids and peptides. It is used as an antifreeze, solvent, fuel, and as a denaturant for ethanol. It is also used for producing biodiesel. It is used for the synthesis of formaldehyde and acetic acid. It is also useful to make chemicals, to remove water from automotive and aviation fuels, as a solvent for paints and plastics, and as an ingredient in a wide variety of products. It plays an important role for the measurement of methyl ester content of pectin.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point °C to 98°C
Color Colorless
Appearance Colorless liquid
Density 0.791
Quantity 4 x 1 L
Flash Point 11°C (52°F)
Odor Mild
Physical Form Liquid
Refractive Index 1.329
Assay Percent Range >99.8%
Vapor Pressure 128 hPa (96mm Hg) at 20°C
Beilstein 1098229
Boiling Point 64.7°C
Solubility Information Miscible with water and most of the organic solvents including ethyl acetate,toluene,dichloromethane,chloroform,tetrahydrofuran,and acetone.
Formula Weight 32.04
Grade ACS
Chemical Name or Material Methanol
Show More Show Less
Hazard Category H225-H301+H311+H331-H370-H372
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation DOT Class: 3; Packing Group: II
EINECSNumber 200-659-6
RTECSNumber PC1400000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.