Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol ACS, MilliporeSigma™
GSA_VA

Catalog No. MMX04857 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
20 L
200 L
Packaging:
Glass Bottle
Metal Can
Metal Drum
Poly Bottle
Poly-coated Glass Bottle
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
MMX04857 4 L Glass Bottle
MMX04856 500 mL Glass Bottle
MMX04858 1 L Glass Bottle
MMX04859 1 L Poly Bottle
MMX04853 4 L Poly Bottle
MMX0485P4 4 L Poly-coated Glass Bottle
MMX04855 20 L Metal Can
MMX048520 200 L Metal Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. MMX04857 Supplier MilliporeSigma Supplier No. US1MX0485/7
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -97.8°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 4 L
Flash Point Closed cup: 12°C (53.6°F), Open cup: 15.85°C (60.5°F)
Ignition Point 464°C (867.2°F)
Odor Alcohol-like
Packaging Glass Bottle
Physical Form Liquid
Linear Formula CH3OH
UN Number UN 1230
Solubility Information Soluble in water
Formula Weight 32.04
Percent Purity ≥99.9%
Grade ACS/NF/USP
Chemical Name or Material Methanol
Show More Show Less
DOTInformation DOT Class: 3, Packaging Group: II
Recommended Storage Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.