Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), Reagents

Catalog No. 01338130
Change view
Click to view available options
Quantity:
4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-338-130 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-338-130 Supplier Reagents Holdings Llc Supplier No. C11210204C1
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

Methanol, ACS Reagent, Absolute (Methyl Alcohol Anhydrous), ACS, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl Alcohol, Wood Alcohol
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Quantity 4 L
Packaging Safety Coated Glass
Physical Form Liquid
Specific Gravity 0.7866
Boiling Point 64.5°C
Grade ACS
Chemical Name or Material Methanol, ACS Reagent (Methyl Alcohol)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.