Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Anhydrous, Reagent Special, ACS, 99.8%, Spectrum™ Chemical
SDP

Catalog No. 18604365 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
500 mL
1 L
4 L
6 x 500 mL
20 L
4 x 4 L
6 x 1 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-365 6 x 1 L Amber Glass Bottle
18-604-361 500 mL Amber Glass Bottle
18-604-363 1 L Amber Glass Bottle
18-604-362 6 x 500 mL Amber Glass Bottle
18-604-364 4 L Amber Glass Bottle
18-604-366 4 x 4 L Amber Glass Bottle
18-604-367 20 L Steel Can
18-604-368 200 L Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
Catalog No. 18-604-365 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M12351LTCS6
Add to Cart
Add to Cart

M1235, 67-56-1, CH3OH

Spectrum™ Chemical Methanol, Anhydrous, Reagent Special, ACS is also known by several other names including methyl alcohol, wood alcohol, and wood spirits. Methanol is known as the simplest alcohol. It is light, extremely volatile, colorless, flammable liquid with a strong odor. Methanol is highly toxic and it is not safe for human consumption. It is commonly used as an antifreeze, solvent, and a fuel as well as a denaturant for ethanol. As an ACS grade quality reagent, its chemical specifications are the de facto standards for chemicals used in many high-purity applications and typically designate the highest quality chemical available for laboratory use. Spectrum Chemical manufactured Reagent ACS grade products meet the toughest regulatory standards for quality and purity.

Chemical Identifiers

Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

CAS 67-56-1
CAS Max % 100%
Quantity 6 x 1 L
Packaging Amber Glass Bottle
Residue after Evaporation 0.001%
Titratable Acid 0.0003 meq/g
Titratable Base 0.0002 meq/g
UN Number UN1230
Boiling Point 64°C to 65°C
Percent Purity 99.8%
Grade Reagent
Chemical Name or Material Methanol
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.