Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Anhydrous, septum-sealed, for applications requiring low water content, >99.9%, Solstice

Catalog No. 6000930 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
6 x 100 mL
Packaging:
Septum-sealed Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-009-30 6 x 100 mL Septum-sealed Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-009-30 Supplier Solstice Supplier No. AS2300100

missing translation for 'validMainframeMsg'

Anhydrous, septum-sealed, for applications requiring low water content, >99.9%

  • Water content ranging from <10 ppm to <50 ppm
  • Low residue
  • Convenient and effective packaging ensures product quality
  • Certificate of Analysis printed on the label
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.0000%
Density 0.792 g/cm3
Quantity 6 x 100 mL
Flash Point 11°C
Packaging Septum-sealed Bottle
Physical Form anhydrous
UN Number UN1230
Beilstein 1098229
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity >99.9%
Chemical Name or Material Methanol, Anhydrous, septum-sealed, for applications requiring low water content, >99.9%
Show More Show Less
ShelfLife 365 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.