Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, BioSyn™, for biosynthesis and applications requiring low water content, >99.9%, Solstice

Catalog No. 60048487 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-048-487 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-487 Supplier Solstice Supplier No. BB2334
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

  • Ultra-low water specifications
  • Low amine specifications
  • Low particulate levels
  • High solvent purity
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl Alcohol, Carbinol, Wood Alcohol
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.00
Density 0.792 g/cm3
Quantity 4 x 4 L
Flash Point 11°C
Packaging Glass Bottle
Physical Form Liquid
Boiling Point 64.7°C
Grade Low Water
Hazard Category Danger
Hazard Statement H225+H370
Precautionary Statement P210+P233+P240+P241+P242+P243+P260+P264+P270+P280
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.