Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol (Certified ACS), Fisher Chemical™ PROMO

Catalog No. A4124S24 Shop All Fisherbrand Products
Change view
Click to view available options
Methanol (ACS); Packaging: Glass Bottle; Quantity: 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
A4124S24 Methanol (ACS); Packaging: Glass Bottle; Quantity: 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A4124S24

Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.

View Alternate Product

Synonym: Methyl Alcohol, Wood Alcohol, Carbinol

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.791 g/cm3
Quantity 4 L
Flash Point 12°C (53.6°F)
Identification Pass Test
Packaging Glass Bottle
Physical Form Liquid
Residue after Evaporation 10 ppm Maximum
Assay Percent Range ≥99.8%
Specific Gravity 0.791
Titratable Acid 0.0003 mEq/g
Titratable Base 0.0002 mEq/g
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55 cP at 20°C
UN Number 1230
Water 0.1%
Solubility Information Soluble in Water
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity 99.8%
Grade ACS
Chemical Name or Material Methanol
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement May cause fire or explosion; strong oxidizer.
Precautionary Statement Toxic if inhaled. Toxic in contact with skin. Toxic if swallowed. Causes skin irritation. Causes eye irritation. May cause respiratory irritation.
DOTInformation DOT Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.