Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, For pesticide residue analysis, Solstice

Catalog No. 60046346 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2.5 L
4 x 2.5 L
4 x 4 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-046-346 4 x 2.5 L
60-009-57 1 L
60-009-58 2.5 L
60-009-63 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 60-046-346 Supplier Solstice Supplier No. 344854X2.5L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™, for pesticide residue analysis

  • For gas chromatography (GC)
  • Developed especially for the application in residue analysis of pesticides and other low-volatile substances using GC
  • Non-volatile matter and water content are specified for each solvent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Density 0.791g/mL at 25°C
Quantity 4 x 2.5 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Linear Formula CH3OH
Vapor Pressure 410 mmHg (50°C), 97.68 mmHg (20°C)
UN Number UN1230
Formula Weight 32.04
Percent Purity ≥99.9% (GC)
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P311
DOTInformation class 3 - PG 2 - Methanol
RTECSNumber 200-659-6
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.