Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol-d{4}, 100%(Isotopic)

Catalog No. AA42320KV
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42320KV 10 Ea.
AA42320SA 2 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42320KV Supplier Thermo Scientific Chemicals Supplier No. 042320KV
Add to Cart

CAS: 811-98-3 | CH4O | 36.066 g/mol

Methanol-d_4 is the labeled analog of methyl alcohol, a common laboratory solvent used primarily in spectroscopic techniques (UV/VIS, NMR, IR).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
Molecular Weight (g/mol) 36.066
MDL Number MFCD00044637
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
PubChem CID 71568
IUPAC Name trideuterio(deuteriooxy)methane
SMILES CO

Specifications

Physical Form Liquid
Color Colorless
Quantity 10 Ea.
Packaging Prescored Ampule
Density 0.89 g/mL
Odor Mild
Flash Point 11°C (52°F)
Melting Point -98°C
Boiling Point 64.7°C
UN Number UN1230
Solubility Information Fully miscible in water.
Formula Weight 36.07
Percent Purity 100%
Chemical Name or Material Methanol-d4, Isotopic
Show More Show Less
missing translation for 'healthHazard1' H225-H301+H311+H331-H370
missing translation for 'healthHazard2' GHS H Statement
H225-H300-H310-H330-H370
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs.
missing translation for 'healthHazard3' P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL
EINECSNumber 212-378-6
TSCA Yes
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.