Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol-d{4}, 99.8% (Isotopic), contains 0.05% v/v TMS

Catalog No. AA42319KV
Change view
Click to view available options
Quantity:
10 Ea.
2 Ea.
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA42319KV 10 Ea.
AA42319SA 2 Ea.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AA42319KV Supplier Thermo Scientific Chemicals Supplier No. 042319KV
Add to Cart

CAS: 811-98-3 | CH4O | 36.066 g/mol

Methyl alcohol-d4 is the labeled analog of methyl alcohol, a common laboratory solvent. It is a common solvent used in NMR spectroscopy.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 811-98-3
Molecular Formula CH4O
Molecular Weight (g/mol) 36.066
MDL Number MFCD00044637
InChI Key OKKJLVBELUTLKV-MZCSYVLQSA-N
Synonym methanol-d4, perdeuteromethanol, tetradeuteromethanol, methan-d3-ol-d, 2h4 methanol, methyl alcohol-d4, cd3od, methyl-d3 alcohol-d, trideuterio deuteriooxy methane, methanol-d4, 99.8 atom % d
PubChem CID 71568
IUPAC Name trideuterio(deuteriooxy)methane
SMILES CO

Specifications

Physical Form Liquid
Packaging Prescored Ampule
Quantity 10 Ea.
Color Colorless
Density 0.89 g/mL
Odor Mild
Melting Point -98°C
Boiling Point 64.7°C
Linear Formula CD3OD
UN Number UN1230
Solubility Information Miscible with water,ethanol,ether,acetone and chloroform.
Formula Weight 36.07
Percent Purity 99.8%
Concentration or Composition (by Analyte or Components) Contains 0.05% v/v TMS
Chemical Name or Material Methanol-d4, Isotopic
Show More Show Less
Hazard Category H225-H301+H311+H331-H370
Hazard Statement GHS H Statement
H225-H300-H310-H330-H370
Highly flammable liquid and vapor.
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P310-P303+P361+P353-P304+P340-P307+P311-P311-P312-P330-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: METHANOL
EINECSNumber 212-378-6
TSCA Yes
Recommended Storage Ambient

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.