Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol For gas chromatography ECD and FID SupraSolv™, MilliporeSigma™

Catalog No. M1060112500 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
2.5 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
M1060112500 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M1060112500 Supplier MilliporeSigma Supplier No. 1.06011.2500
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For gas chromatography ECD and FID SupraSolv™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.79g/cm3 at 20°C
Quantity 2.5 L
Flash Point 10°C (50°F)
Ignition Point 455°C (851°F)
Odor Charactristic
Packaging Glass Bottle
Vapor Pressure 128 hPa (20°C)
UN Number 1230
Boiling Point 64.5°C
Solubility Information Soluble in water at 20°C
Formula Weight 32.04
Chemical Name or Material Methanol
Show More Show Less
Hazard Category H225: Highly flammable liquid and vapour.
Hazard Statement H301 + H311 + H331: Toxic if swallowed, in contact with skin or if inhaled
Precautionary Statement H370: Causes damage to organs
Recommended Storage 5°C to 30°C (41°F to 86°F); Keep container tightly closed in a dry and well-ventilated place. Keep away from heat and sources of ignition. Keep locked up or in an area accessible only to qualified or authorized persons.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.