Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, For HPLC analysis, J.T. Baker™

Catalog No. 02004653 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Narrow Mouth Amber Glass Bottle
Narrow Mouth PVC-coated Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
02-004-653 4 L Narrow Mouth Amber Glass Bottle
02-003-342 1 L Narrow Mouth Amber Glass Bottle
02-003-349 4 L Narrow Mouth PVC-coated Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 02-004-653 Supplier Avantor Supplier No. 909303
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High purity solvents for use in analytical and preparative separations

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -97.6°C
Color Colorless
Density 0.7918g/cm3
Quantity 4 L
Flash Point 12°C
Packaging Narrow Mouth Amber Glass Bottle
Physical Form Liquid
Specific Gravity 1L = 0.80kg
Boiling Point 64.7°C
Formula Weight 32.04g/mol
Grade HPLC
Chemical Name or Material Methanol, For HPLC analysis
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.