Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, For HPLC, Gradient Grade, Suitable as ACS-grade LC Reagent, ≥99.9%, MilliporeSigma™ Supelco™

Catalog No. 111005330 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
50 L
Packaging:
Pure-Pac™ 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
11-100-5330 50 L Pure-Pac™ 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11-100-5330 Supplier MilliporeSigma Supelco Supplier No. 43919350LP24A
Add to Cart
Add to Cart

Methanol (MeOH) is an organic solvent with a wide range of industrial applications. It is the main component of lithium batteries. It can be prepared by hydrogenation of CO2 under various conditions. MeOH is a toxic alcohol employed for denaturing ethyl alcohol. A study reports the preparation of biodiesel from canola oil using methanol under supercritical conditions.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym Methyl alcohol
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color APHA: ≤10
Density 0.791 g/mL (at 25°C)
Quantity 50 L
Packaging Pure-Pac™ 2
Refractive Index n20/D 1.329
Linear Formula CH3OH
Vapor Pressure 410 mmHg (50°C)
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity ≥99.9%
Grade HPLC Gradient
Show More Show Less
RTECSNumber PC1400000
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.