Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Solstice

Catalog No. 60090373 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
200 L
Packaging:
Returnable Stainless Steel Drum
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-090-373 200 L Returnable Stainless Steel Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-090-373 Supplier Solstice Supplier No. CS230ZC200
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

ACS Reagent, meets ACS Specifications for General Use, >99.8%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
PubChem CID 887
ChEBI CHEBI:17790

Specifications

Quantity 200 L
Flash Point 11 °C
Packaging Returnable Stainless Steel Drum
Physical Form Liquid, Clear
Grade ACS
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.