Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Solstice

Catalog No. 60090378 Shop All Solstice Research Chemicals Products
Catalog No. 60-090-378 Supplier Solstice Supplier No. BC23356
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

BioSyn™, for biosynthesis and applications requiring low water content, >99.9%

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
PubChem CID 887
ChEBI CHEBI:17790

Specifications

Flash Point 11 °C
Packaging Returnable Steel Drum
Physical Form Liquid, Clear
Molecular Formula CH4O
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Molecular Weight (g/mol) 32.04
Recommended Storage Room Temperature
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.