Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl Alcohol, MP Biomedicals™

Catalog No. ICN30014116 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 L
4 L
4 x 4 L
6 x 1 L
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN30014116 4 x 4 L
ICN30014104 4 L
ICN30014106 6 x 1 L
ICN30014101 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. ICN30014116 Supplier MP Biomedicals, Inc Supplier No. 0230014116
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methyl Alcohol is used as industrial solvent.

  • Starting material for organic synthesis of formaldehydes, celluloid, textile soap, wood stains, coated fabrics, shatter-proof glass, paper coating, waterproofing formulations, artificial leather, synthetic indigo and other dyes.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
IUPAC Name methanol

Specifications

Quantity 4 x 4 L
Formula Weight 32.04
WARNING: Reproductive Harm - www.P65Warnings.ca.gov

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.