Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, HPLC Grade, Inventus Biotech
SDP

Catalog No. 21520138
Change view
Click to view available options
Quantity:
4 x 4 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
21-520-138 4 x 4 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 21-520-138 Supplier Cen-Med Enterprises Supplier No. 393326HPLC
Add to Cart
Add to Cart

HPLC Grade Methanol is a high purity solvent (> 99.9%), ideally suited for HPLC applications and general laboratory use.

  • HPLC Grade Methanol is 0.2 micron filtered, and also exceeds all ACS specifications
  • Ultra Purified
  • Sold in Amber Glass Bottles
  • Case of 4 x 4L

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 887
ChEBI 17790
IUPAC Name methanol
SMILES CO

Specifications

Quantity 4 x 4 L
Identification Methanol, Methyl alcohol, Wood alcohol
Packaging Amber Glass Bottle
Linear Formula CH3OH
UN Number 1230
Formula Weight 32.042
Grade HPLC
Chemical Name or Material Methanol, For HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.