Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, HPLC Plus, ≥99.9%, Solstice

Catalog No. 60048242 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
4 x 4 L
Packaging:
Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-048-242 4 x 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 60-048-242 Supplier Solstice Supplier No. 6463774X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CHROMASOLV™ Plus, for HPLC

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Density 0.791g/mL at 25°C, 0.791g/mL at 25°C
Quantity 4 x 4 L
Explosion Limit 0.36
Ignition Point 725°C
Packaging Glass Bottle
Vapor Pressure 410mmHg (50°C), 97.68mmHg (20°C)
Linear Formula CH3OH
UN Number UN1230
Beilstein 1098229
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha
Percent Purity ≥99.9%
Grade HPLC Plus
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P311
DOTInformation class 3 - PG 2 - Methanol
RTECSNumber 200-659-6
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.