Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Laboratory Reagent, ≥99.6%, Solstice

Catalog No. 60045009 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
4 x 2.5 L
4 x 4 L
Packaging:
Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-045-009 4 x 4 L Glass Bottle
60-045-005 2.5 L Glass Bottle
60-045-008 4 x 2.5 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 60-045-009 Supplier Solstice Supplier No. 1799574X4L
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Laboratory Reagent,≥99.6%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Density 0.791g/mL at 25°C
Quantity 4 x 4 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Packaging Glass Bottle
Physical Form Liquid
Refractive Index n20/D 1.329(lit.)
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
UN Number UN1230
Beilstein 1098229
Boiling Point 65°C
Formula Weight 32.04g/mol
Percent Purity ≥99.6%
Grade Laboratory Reagent
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement P210-P280-P302 + P352 + P312-P304 + P340 + P312-P370 + P378-P403 + P235
Precautionary Statement H225-H301 + H311 + H331-H370
DOTInformation class 3 - PG 2 - Methanol
EINECSNumber 200-659-6
RTECSNumber PC1400000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.