Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, LC-MS Ultra CHROMASOLV™, Solstice
GSA_VA

Catalog No. p-7071822 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
1 L
2 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
60-009-64 1 L
60-009-65 2 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-009-64 Supplier Solstice Supplier No. 142621L
Only null left
Add to Cart
Add to Cart

CHROMASOLV™ LC-MS Ultra, tested for UHPLC-MS

  • For the most demanding LC-MS and UHPLC applications
  • Tested for UHPLC-MS suitability
  • Highest solvent purity for low detection limits
  • Extremely low MS baseline noise
  • Exemplary lot-to-lot reproducibility
  • Low levels of trace metals
  • Packaged in clear borosilicate glass bottles

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Density 0.791g/mL at 25°C
Quantity 1 L
Explosion Limit 0.36
Flash Point 9°C (48.2°F)
Ignition Point 725°F
Linear Formula CH3OH
Vapor Pressure 97.68mmHg (20°C), 410mmHg (50°C)
UN Number 1230
Beilstein 1098229
Formula Weight 32.04g/mol
Grade LC-MS Ultra CHROMASOLV
Chemical Name or Material Methanol
Show More Show Less
Hazard Statement P210-P260-P280-P301 + P310-P311
Precautionary Statement H225-H301 + H311 + H331-H370
EINECSNumber 200-659-6
RTECSNumber PC1400000
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.