Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol LM Prime, Transene

Catalog No. 502744716
Encompass
Catalog No. 50-274-4716 Supplier Thermo Scientific Production Chemicals and Services Supplier No. T0701355GAL
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methanol Semiconductor Grade

Specifically formulated for electronics manufacturing are referred to as electronic grade solvents. These solvents contain very low levels of heavy metals, typically measured in parts per billion or parts per trillion. Achieving the required purity levels involves several stages of purification. They are commonly utilized as cleaners and etchants in the production of semiconductors and microprocessors.

  • Common Uses and Applications:
  • HPLC (High-Performance Liquid Chromatography)
  • Reagent
  • Electronics
  • Cleaning agent
  • Semiconductor synthesis
  • Solvent
  • Etching agent

Safety and Shipping Information:DOT: Methanol, 3, UN1230, PG II

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Weight (g/mol) 32.04

Specifications

Packaging HDPE
Molecular Formula CH4O
Boiling Point 64.7°C
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
SMILES CO
IUPAC Name methanol
Grade Electronic
Chemical Name or Material Methyl Alcohol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.