Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, MilliporeSigma

Catalog No. 1037262002 Shop All MilliporeSigma Products
Catalog No. 10-372-62002 Supplier MilliporeSigma Supplier No. 1037262002
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

UHPLC-MS LiChrosolv™ solvents are designed to meet the highest requirements of UHPLC-MS in research and quality control, including proteomics and metabolomics and environmental, clinical, food or industrial testing applications.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
IUPAC Name methanol
SMILES CO

Specifications

Packaging Borosilicate Glass Bottle
Boiling Point 64.7°C
Chemical Name or Material Methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.