Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol (Peroxide-Free/Sequencing), Fisher BioReagents™

Catalog No. BP11051 Shop All Fisherbrand Products
Change view
Click to view available options
Quantity:
1 L
4 L
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
BP1105-1 1 L Glass Bottle
BP1105-4 4 L Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BP1105-1

Methanol (Peroxide-Free/Sequencing)

Methanol is used in staining and destaining protein electrophoresis gels, in HPLC, and in other biological applications.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Absorbance 0.025 max. at 254nm, 0.15 max. at 230nm, 0.3 max. at 220nm, 1 max. at 205nm
Color Colorless
Density 0.791g/cm³
Quantity 1 L
Packaging Glass Bottle
Physical Form Liquid
Refractive Index 1.326 to 1.330 (at 25°C)
Residue after Evaporation 3ppm max.
Assay Percent Range ≥99.9 %
Titratable Acid 0.0003mEq/g max.
Linear Formula CH3OH
Titratable Base 0.0002mEq/g max.
Vapor Pressure 128 hPa at 20°C
Viscosity 0.55 cP at 20°C
Water 0.1% max.
Merck Index 15, 6029
Boiling Point 64.7°C
Formula Weight 32.04
Percent Purity ≥99.9%
Grade Peroxides: ≤0001%
Chemical Name or Material Methanol, Peroxide-free
Show More Show Less
Safety handling Solvent Safety handling Check Alert storage code Red
Hazard Statement DANGER!
Precautionary Statement Emergency Overview
Flammable liquid and vapor. Cannot be made non-poisonous. Vapor harmful. Toxic by inhalation, in contact with skin and if swallowed. Irritating to eyes and skin. May cause irritation of respiratory tract. Toxic: danger of very serious irreversible effects through inhalation, in contact with skin and if s. Explosive. Use personal protective equipment. Keep away from open flames, hot surfaces and sources of ignition. Use only under a chemical fume hood. Wash off immediately with plenty of water for at least 15 minutes. Wash off immediately with plenty of water for at least 15 minutes. Immediate medical attention is required.

NFPA

Health:1
Flammability:3
Instability:0
DOTInformation DOT Class 3, : Flammable Liquid
Recommended Storage RT
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.