Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Purge and Trap, ULTRA RESI-ANALYZED, J.T. Baker™

Catalog No. 02004652 Shop All Avantor Products
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-004-652 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 02-004-652 Supplier Avantor Supplier No. 907702
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

High purity solvents and salts developed for organic residue extraction/concentration procedures

A group of ultra high purity solvents and salts developed for organic residue extraction/concentration procedures.

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

Melting Point -98°C
Color Colorless
Quantity 1 L
Explosion Limit 5.5 to 44 vol%
Flash Point 11°C
Ignition Point 455°C
Odor Alcohol-like
Physical Form Liquid
Titratable Acid ≤0.3μeq/g
UN Number 1230
Solubility Information Fully miscible in water
Boiling Point 64.7°C
Percent Purity ≥99.9%
Grade ULTRA RESI-ANALYZED
Chemical Name or Material Methanol, Purge and Trap
Show More Show Less
Safety handling Check Alert storage code Red
Hazard Category Danger
Hazard Statement Highly flammable liquid and vapour. Toxic if swallowed. Toxic in contact with skin.Toxic if inhaled. Causes damage to organs
Precautionary Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower. Store locked up. Dispose of contents/container in accordance with local/regional/national/international regulations.
DOTInformation Class 3: Flammable Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.