Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methanol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9%, Solstice

Catalog No. 6000983 Shop All Solstice Research Chemicals Products
Change view
Click to view available options
Quantity:
19 L
204 L
Packaging:
Metal Can
Metal Drum
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
60-009-83 204 L Metal Drum
60-009-82 19 L Metal Can
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 60-009-83 Supplier Solstice Supplier No. PP230204
Add to Cart
Add to Cart
Request a Quote

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 67-56-1
Molecular Formula CH4O
Molecular Weight (g/mol) 32.04
MDL Number MFCD00004595
InChI Key OKKJLVBELUTLKV-UHFFFAOYSA-N
Synonym methyl alcohol, wood alcohol, carbinol, wood spirit, wood naphtha, methylol, methyl hydroxide, pyroxylic spirit, colonial spirit, columbian spirit
PubChem CID 887
ChEBI CHEBI:17790
IUPAC Name methanol
SMILES CO

Specifications

CAS Max % 100.0000%
Density 0.792 g/cm3
Quantity 204 L
Flash Point 11°C
Packaging Metal Drum
Water 0.1 % max.
UN Number UN1230
Beilstein 1098229
Boiling Point 64.7°C
Solubility Information Miscible in Water at 20°C
Formula Weight 32.04
Percent Purity >99.9%
Grade ACS
Assay by GC Analysis
Chemical Name or Material Methanol, Purified Plus™, ACS Reagent, for organic synthesis, for prep-LC, >99.9%
Show More Show Less
ShelfLife 547 days from date of manufacture
DOTInformation Transport Hazard Class: 3; Packing Group:3: II; Proper Shipping Name: Methanol
EINECSNumber 200-659-6
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.