Learn More
Thermo Scientific Chemicals Betamethasone, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Loss on Drying | 0.5% max. |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 96% min. (HPLC) |
| Quantity | 5 g |
| Solubility Information | Solubility in water: slightly soluble. |
| Formula Weight | 392.46 |
| Percent Purity | 97% |
| Chemical Name or Material | βmethasone |
Safety and Handling
| Hazard Category | GHS Signal Word: Danger |
| Hazard Statement | GHS H Statement May damage the unborn child. May cause damage to organs through prolonged or repeated exposure. |
| Precautionary Statement | GHS P Statement Obtain special instructions before use. Use personal protective equipment as required. IF exposed or concerned: Get medical advice/attention. Do not breathe dust/fume/gas/mist/vapors/spray. |
| EINECSNumber | 206-825-4 |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.