Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ MethElute™ Reagent (TMPAH)

Catalog No. PI49301 Shop All Thermo Scientific Products
Encompass
Change view
Click to view available options
Quantity:
12 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI49301 12 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI49301 Supplier Thermo Scientific™ Supplier No. TS49301
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

For accurate, sensitive on-column methylation use Thermo Scientific™ MethElute™ Reagent (TMPAH).

For accurate, sensitive, on-column methylation and detection by GC analysis use Thermo Scientific™ MethElute™ Reagent (TMPAH). It is ideally suited for the quantitative methylation and detection of barbiturates, sedatives, xanthine bases, phenolic alkaloids, and phenytoin, giving a single quantitative peak for each substance. When the reagent is heated with drug-containing extracts, serum, or urine the drugs containing reactive amino, hydroxyl, and carboxyl functions will be methylated at the reactive sites.

Performance Characteristics

Accuracy comparable or better than the UV/TLC method (GC is superior to UV/TLC when phenobarbitol and phenytoin present)

High precision, with coefficient of variation of 5% or less

High sensitivity; barbiturates deteced down to 0.2 mg/dL

TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Hazards Flammable, Toxic
Standard Components Trimethylanilinium hydroxide in Methanol
Standard Concentration 0.2 M
Quantity 12 x 1 mL
Storage Room Temperature

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.