Learn More
Description
Methimazole USP is a white to pale buff, crystalline powder which has a faint characteristic odor.
Specifications
Specifications
| Chemical Name or Material | Methimazole USP |
| Packaging | Jar 300ml, White, HDPE, Round, 53-400, TRIVENI-DB, CT |
| CAS | 60-56-0 |
| Type | Drug Standard |
| Molecular Weight (g/mol) | 114.17 |
| Quantity | 100 g |
| Percent Purity | 98 to 101 % |
| Grade | USP |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
