Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methocarbamol, USP, 98.5-101.5%, Spectrum™ Chemical
SDP

Catalog No. 18604390 Shop All Spectrum Chemical Manufacturing Corporation Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
1 kg
2.5 kg
25 kg
Packaging:
Amber Glass Bottle
Fiber Drum
Poly Pail
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18-604-390 100 g Amber Glass Bottle
18-604-389 25 g Amber Glass Bottle
18-604-391 500 g Amber Glass Bottle
18-604-392 1 kg Amber Glass Bottle
18-604-394 2.5 kg Poly Pail
18-604-393 25 kg Fiber Drum
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. 18-604-390 Supplier Spectrum Chemical Manufacturing Corporation Supplier No. M1088100GM
Add to Cart
Add to Cart

M1088, 532-03-6, C11H15NO5

Spectrum™ Chemical Methocarbamol, USP is used to treat muscle spasms and pain. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 532-03-6
Molecular Formula C11H15NO5
Molecular Weight (g/mol) 241.24
InChI Key GNXFOGHNGIVQEH-UHFFFAOYNA-N
IUPAC Name 2-hydroxy-3-(2-methoxyphenoxy)propyl carbamate
SMILES COC1=CC=CC=C1OCC(O)COC(N)=O

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 0.5%
Packaging Amber Glass Bottle
Quantity 100 g
Percent Purity 98.5 to 101.5%
Grade USP
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.