Learn More
Microsolv Technology Corporation Method Development Kit 2104, TYPE-C. Includes 5 Cogent HPLC Columns, 2.1mm x 100mm. 1 PK.
Supplier:  Microsolv Technology Corporation 4010010P2
Encompass_Preferred
This Kit, 2104 is an economical way to purchase 5 Cogent HPLC Columns that are often used with LCMS for Method Development and Scouting. Each Column has a 2.1mm Internal Diameter and is 100mm Long, packed with 4um particles, 100A pore size and are manufactured with Cogent TYPE-C™ Silica Hydride. This kit includes Cogent Diamond Hydride™, Cogent Bidentate C18™, Cogent Bidentate C8™, Cogent Silica-C™, Cogent UDC-Cholesterol™.
Catalog No. 50-227-0814
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.