Learn More
R&D Systems Method Development Kit
SDP
Catalog No. NC0736898 Shop All Novus Products
Encompass
This kit includes a test mix, anolyte, catholyte, methyl cellulose, five types of ampholytes (Pharmalyte pH ranges 3-10, 2.5-5, 5-8 and 8-10.5 and Servalyt pH range 2-9), eight pI markers (3.21, 5.12, 6.14, 7.05, 7.65, 8.18, 9.46 and 10.10) and additives (urea and arginine).
Catalog No. NC0736898 Supplier R&D Systems Supplier No. 042848/DEL
Customers require a valid business account associated to their profile to access pricing and ordering options.
This kit includes a test mix, anolyte, catholyte, methyl cellulose, five types of ampholytes (Pharmalyte pH ranges 3-10, 2.5-5, 5-8 and 8-10.5 and Servalyt pH range 2-9), eight pI markers (3.21, 5.12, 6.14, 7.05, 7.65, 8.18, 9.46 and 10.10) and additives (urea and arginine).