Learn More
Thermo Scientific™ Method Specific Reagents for EliA™ Tests (Phadia™ 2500/5000)
Catalog No. 83101701P
Change view
Click to view available options
Type:
EliA IgA Calibrator Strips
EliA IgA Calibrator Wells
EliA IgA Curve Control Strips
EliA IgG Calibrator Strips
EliA IgG Curve Control Strips
EliA IgM Calibrator Strips
EliA IgM Calibrator Wells
EliA IgM Curve Control Strips
EliA™ IgA Conjugate 200
EliA™ IgA Conjugate 50
EliA™ IgG Conjugate 200
EliA™ IgG Conjugate 50
Quantity:
1200 Tests
300 Tests
48 Tests
6 Vials
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Type Quantity
83-101-701P EliA™ IgG Conjugate 50 300 Tests
14-552-701P EliA IgM Calibrator Wells 48 Tests
14-551-601P EliA IgA Calibrator Wells 48 Tests
83-101-601P EliA IgG Curve Control Strips 6 Vials
83-101-501P EliA IgG Calibrator Strips 6 Vials
83-102-001P EliA IgA Curve Control Strips 6 Vials
83-101-901P EliA IgA Calibrator Strips 6 Vials
83-102-101P EliA™ IgA Conjugate 50 300 Tests
83-101-801P EliA™ IgG Conjugate 200 1200 Tests
83-102-201P EliA™ IgA Conjugate 200 1200 Tests
83-105-201P EliA IgM Calibrator Strips 6 Vials
83-105-301P EliA IgM Curve Control Strips 6 Vials
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. 83-101-701P Supplier Thermo Scientific™ Supplier No. 83101701
Add to Cart
Add to Cart
Reagents support EliA, an enzyme immunoassay to measure autoantibodies for the diagnosis of autoimmune diseases such as rheumatoid arthritis, celiac disease or systemic lupus erythematosus.
TRUSTED_SUSTAINABILITY
Specifications
Detectable Analytes Autoimmune Disease Autoantibodies
Type EliA™ IgG Conjugate 50
For Use With (Application) EliA™ Tests (Phadia™ 2500/5000)
Quantity 300 Tests