Learn More
Lkt Laboratories CAS 6745-93-3, ≥98%, DHF reductase inhibitor used to treat rheumatoid arthritis, ectopic pregnancy, and various cancers. It prevents synthesis of thymidine, RNA, and RNA.
SDP
Supplier:  Lkt Laboratories M1676
Encompass_Preferred
Methotrexate is an antimetabolite that exhibits immunosuppressive, abortifacient, anti-inflammatory, and anticancer chemotherapeutic activities. Methotrexate is clinically used to treat various cancers, ectopic pregnancy, and rheumatoid arthritis. Methotrexate inhibits dihydrofolate reductase, preventing thymidine, DNA, and RNA synthesis. In vivo, methotrexate increases T cell apoptosis, decreases T cell activation, and suppresses cytokine production.
Catalog No. NC2530040
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.