Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxy(dimethyl)octadecylsilane 90.0+%, TCI America™
SDP

Catalog No. M301125ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M301125ML 25 mL
M30115ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M301125ML Supplier TCI America Supplier No. M301125ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 71808-65-6
Molecular Formula C21H46OSi
Molecular Weight (g/mol) 342.68
MDL Number MFCD00042920
InChI Key RUFRLNPHRPYBLF-UHFFFAOYSA-N
PubChem CID 172903
IUPAC Name methoxydimethyloctadecylsilane
SMILES CCCCCCCCCCCCCCCCCC[Si](C)(C)OC

Specifications

Color Yellow
Quantity 25 mL
Formula Weight 342.68
Percent Purity ≥90.0% (GC)
Physical Form Liquid
Chemical Name or Material Methoxy(dimethyl)octadecylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.