Learn More
Abcam Methoxy-X04, Amyloid Beta Fluorescent Marker, >98% Purity, 25 mg
Description
- Molecular Weight: 344.4 Da
- Purity: >98%
- Properties: Blood-brain barrier permeable, blue fluorophore
- Function: Selectively binds fibrillar β-sheet deposits with high affinity (Ki = 26.8 nM)
- Applications: Detects plaques, tangles and cerebrovascular amyloid in vivo
- Derivatives: Derivative of Congo Red (ab145645) and Chrysamine G
- CAS Number: 863918-78-9
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Target Molecule | Amyloid Beta |
| Substrate | Amyloid Beta |
| Substrate Properties | Chemical Substrate |
| Shipping Condition | Ambient |
| Label or Dye | Fluorescent Marker |
| Detection Location | In-Vivo Detection |
| Buffer | 100% Methoxy-X04 |
| Content And Storage | Store at 4°C. |
| Format | Vial |
| Detection Method | Fluorescence |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.