Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyacetaldehyde Diethyl Acetal 97%, Thermo Scientific™

Catalog No. AC165160100
Change view
Click to view available options
Quantity:
10 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC165160100 10 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC165160100 Supplier Thermo Scientific Chemicals Supplier No. 165160100

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4819-75-4
Molecular Formula C7H16O3
Molecular Weight (g/mol) 148.202
InChI Key PCYADPMXIILFTP-UHFFFAOYSA-N
Synonym methoxyacetaldehyde diethyl acetal, methoxyacetaldehyde-diethylacetal, ethane, 1,1-diethoxy-2-methoxy, acmc-20akk7, 1,1-diethoxy-2-methoxy-ethane, pcyadpmxiilftp-uhfffaoysa
PubChem CID 78539
IUPAC Name 1,1-diethoxy-2-methoxyethane
SMILES CCOC(COC)OCC

Specifications

Color Colorless
Density 0.91
Infrared Spectrum Authentic
Refractive Index 1.3980 to 1.4000 (20°C, 589nm)
Linear Formula CH3OCH2CH(OC2H5)2
Quantity 10 mL
Formula Weight 148.2
Percent Purity ≥96.0%
Physical Form Liquid
Chemical Name or Material Methoxyacetaldehyde diethyl acetal, 97%
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.