Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyacetone, 96%, Thermo Scientific™

Catalog No. AC175730250
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC175730250 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AC175730250 Supplier Thermo Scientific Chemicals Supplier No. 175730250

missing translation for 'validMainframeMsg'

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5878-19-3
Molecular Formula C4H8O2
Molecular Weight (g/mol) 88.11
MDL Number MFCD00008768
InChI Key CUZLJOLBIRPEFB-UHFFFAOYSA-N
Synonym methoxyacetone, methoxy-2-propanone, 2-propanone, 1-methoxy, 1-methoxy-2-propanone, 1-methoxyacetone, methoxyaceton, methoxymethyl methyl ketone, methoxy acetone, ch3coch2och3
PubChem CID 22172
IUPAC Name 1-methoxypropan-2-one
SMILES COCC(C)=O

Specifications

Density 0.9500g/mL
Boiling Point 118.0°C
Flash Point 25°C
Refractive Index 1.3970 to 1.3990
Linear Formula CH3OCH2COCH3
Quantity 25 g
Specific Gravity 0.95
Solubility Information Solubility in water: miscible.
Formula Weight 88.11
Chemical Name or Material Methoxyacetone
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.