Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyamine Hydrochloride, MP Biomedicals™

Catalog No. ICN15540505 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15540505 5 g
ICN15540501 1 g
ICN15540525 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15540505 Supplier MP Biomedicals, Inc Supplier No. 0215540505
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Methoxyamine Hydrochloride is used as a reagent for the preparation of O-methyl oximes.

  • Also used to make O-methyl oximes from aldehydes, or ketones.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 593-56-6
Molecular Formula CH6ClNO
Molecular Weight (g/mol) 83.515
InChI Key XNXVOSBNFZWHBV-UHFFFAOYSA-N
Synonym o-methylhydroxylamine hydrochloride, methoxyamine hydrochloride, methoxylamine hydrochloride, methoxyammonium chloride, o-methylhydroxylamine hcl, aminooxy methane hydrochloride, methoxyamine hcl, unii-203546olmf, hydroxylamine, o-methyl-, hydrochloride, o-methylhydroxyamine hydrochloride
PubChem CID 521874
IUPAC Name O-methylhydroxylamine;hydrochloride
SMILES CON.Cl

Specifications

Melting Point 300°F to 304°F (decomposes)
Quantity 5 g
Solubility Information Soluble
Formula Weight 83.52
Physical Form Crystalline Powder
Chemical Name or Material Methoxyamine Hydrochloride
RTECSNumber NC3980000
Recommended Storage Room Temperature

Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use. For more information, please contact our customer service department.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.