Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxychlor Solution, 100μg/mL in methanol, Ultra Scientific

Catalog No. USPP3901
Encompass
Change view
Click to view available options
Quantity:
1 x 1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
US-PP-3901 1 x 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. US-PP-3901 Supplier Agilent Technologies Supplier No. PP3901
May include imposed supplier surcharges.
Add to Cart
Add to Cart

Chemical Identifiers

CAS 72-43-5 , 67-56-1
Molecular Formula C16H15Cl3O2
Molecular Weight (g/mol) 345.644
InChI Key IAKOZHOLGAGEJT-UHFFFAOYSA-N
PubChem CID 4115
ChEBI CHEBI:6842
IUPAC Name 1-methoxy-4-[2,2,2-trichloro-1-(4-methoxyphenyl)ethyl]benzene
SMILES COC1=CC=C(C=C1)C(C2=CC=C(C=C2)OC)C(Cl)(Cl)Cl

Specifications

CAS Max % 99.99, 0.01
Analyte or Component Names Methanol,Methoxychlor
Element Methanol,Methoxychlor
Solvent or Matrix Methanol
Quantity 1 x 1 mL
Concentration or Composition (by Analyte or Components) 100μg/mL
Chemical Name or Material Methoxychlor Solution
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.