Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyethanol, 99.3% min., ACS, MilliporeSigma™

Catalog No. MMX05156 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
500 mL
3.1 L
55 gal.
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MMX05156 500 mL
MMX05151 3.1 L
MMX051530 55 gal.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. MMX05156 Supplier MilliporeSigma Supplier No. MX05156
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 109-86-4
Molecular Formula C3H8O2
Molecular Weight (g/mol) 76.095
InChI Key XNWFRZJHXBZDAG-UHFFFAOYSA-N
Synonym methyl cellosolve, ethanol, 2-methoxy, ethylene glycol monomethyl ether, methyl oxitol, 2-methoxy-1-ethanol, methoxyethanol, 3-oxa-1-butanol, egme, monomethyl glycol, dowanol em
PubChem CID 8019
ChEBI CHEBI:46790
IUPAC Name 2-methoxyethanol
SMILES COCCO

Specifications

Color Colorless
Density 0.964g/cm3 (20°C)
Explosion Limit 2.5 to 20%(V)
Ignition Point 325°C
Assay Percent Range 99.3% min.
Linear Formula CH3OCH2CH2OH
Quantity 500 mL
Vapor Pressure 10 hPa (20°C)
Formula Weight 76.9
Grade ACS/EP/Reagent
Physical Form Liquid
Chemical Name or Material 2-Methoxyethanol, For ACS analysis
Show More Show Less
Recommended Storage Store at +15°C to +25°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.