Learn More
Medchemexpress LLC HY-117386 50mg Medchemexpress, Methoxyfenozide CAS:161050-58-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11738650MG
Encompass_Preferred
Medchemexpress, HY-117386 50mg Methoxyfenozide CAS:161050-58-4 Methoxyfenozide, a diacylhydrazine insecticide, selectively binds to lepidopteran ecdysone receptors (EcRs) over dipteran EcRs with Kd values of 0.5 and 124 nM, respectively. Methoxyfenozide is lethal to neonatal larvae of S. exigua, S. frugiperda, T. ni, O. nubilalis, L. pomonella, H. zea, and H. virescens (LC50s=0.35, 0.2, 0.11, 0.18, 0.21, 0.79, and 3.12 mg/L, respectively). Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0903
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.