Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxymethyltrimethylsilane 95.0+%, TCI America™
SDP

Catalog No. M12645ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M12645ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. M12645ML Supplier TCI America Supplier No. M12645ML
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 14704-14-4
Molecular Formula C5H14OSi
Molecular Weight (g/mol) 118.25
MDL Number MFCD00009843
InChI Key PPNQXAYCPNTVHH-UHFFFAOYSA-N
Synonym Trimethylsilylmethyl Methyl Ether
PubChem CID 139809
IUPAC Name (methoxymethyl)trimethylsilane
SMILES COC[Si](C)(C)C

Specifications

Color Colorless
Boiling Point 85°C
Quantity 5 mL
UN Number 3271
Formula Weight 118.25
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Methoxymethyltrimethylsilane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.