Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methoxyphenamine Hydrochloride, MP Biomedicals™

Catalog No. ICN15542905 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15542905 5 g
ICN15542901 1 g
ICN15542925 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. ICN15542905 Supplier MP Biomedicals, Inc Supplier No. 0215542905

Soluble in water (50mg/mL)

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10-3-5588
Molecular Formula C11H18ClNO
Molecular Weight (g/mol) 215.72
MDL Number MFCD00034832
InChI Key WJVCROKJTIUCAZ-UHFFFAOYNA-N

Specifications

Color White
Quantity 5 g
Synonym dl-serine methyl ester hydrochloride, h-dl-ser-ome.hcl, methyl 2-amino-3-hydroxypropanoate hydrochloride, l-serine, hydrochloride, methyl dl-serinate hcl, d,l-serine methyl ester hydrochloride, zlchem 219, dl-serinemethylesterhydrochloride, h-ser-ome??hcl, pubchem10908
SMILES [H+].[Cl-].CCC(NC)C1=CC=CC=C1OC
IUPAC Name hydrogen [1-(2-methoxyphenyl)propyl](methyl)amine chloride
PubChem CID 517373
Formula Weight 215.7
Physical Form Powder
Chemical Name or Material Methoxyphenamine Hydrochloride
EINECSNumber 226-993-2
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.