Learn More
Sigma Organic Chemistry methYdodecYbenzY trime | 250mg | MFCD00171356
Supplier: Sigma Organic Chemistry CDS003485250MG

methYdodecYbenzY trime , 250mg
About this Item:
MDL #: MFCD00171356
Molecular Weight: 368.04
UNSPSC Code: 12352200
Chemical Formula: C23H42ClN
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.