Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 1-Methyl-4-piperidinecarboxylate 98.0+%, TCI America™
SDP

Catalog No. M29611G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M29611G 1 g
M29615G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M29611G Supplier TCI America Supplier No. M29611G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 1690-75-1
Molecular Formula C8H15NO2
Molecular Weight (g/mol) 157.213
MDL Number MFCD06658483
InChI Key KYAOKPRJTMFBTQ-UHFFFAOYSA-N
Synonym 1-Methyl-4-piperidinecarboxylic Acid Methyl Ester
PubChem CID 3616844
IUPAC Name methyl 1-methylpiperidine-4-carboxylate
SMILES CN1CCC(CC1)C(=O)OC

Specifications

Color Yellow
Boiling Point 202°C
Quantity 1 g
Formula Weight 157.21
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Methyl 1-Methyl-4-piperidinecarboxylate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.