Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Methyl 2-[4-(Bromomethyl)phenyl]benzoate 98.0+%, TCI America™
SDP

Catalog No. M180625G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
M180625G 25 g
M18065G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. M180625G Supplier TCI America Supplier No. M180625G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 114772-38-2
Molecular Formula C15H13BrO2
Molecular Weight (g/mol) 305.171
MDL Number MFCD06200816
InChI Key RMXGTMRDXKUUDJ-UHFFFAOYSA-N
PubChem CID 11012181
IUPAC Name methyl 2-[4-(bromomethyl)phenyl]benzoate
SMILES COC(=O)C1=CC=CC=C1C2=CC=C(C=C2)CBr

Specifications

Melting Point 56°C
Color White-Yellow
Quantity 25 g
Synonym 2-[4-(Bromomethyl)phenyl]benzoic Acid Methyl Ester, Methyl 4′-(Bromomethyl)biphenyl-2-carboxylate, 4′-(Bromomethyl)biphenyl-2-carboxylic Acid Methyl Ester
Formula Weight 305.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Methyl 2-[4-(Bromomethyl)phenyl]benzoate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.