Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ Methyl 2,4-dihydroxybenzoate, 97%

Click to view available options
Quantity:
5g
25g
100g
2150-47-2
C8H8O4
168.148
MFCD00002276
IIFCLXHRIYTHPV-UHFFFAOYSA-N
2,4-dihydroxybenzoic acid methyl ester,benzoic acid, 2,4-dihydroxy-, methyl ester,methyl beta-resorcylate,methyl-2,4-dihydroxybenzoate,unii-xe68bu3oyj,xe68bu3oyj,.beta.-resorcylic acid, methyl ester,beta-resorcylic acid methyl ester,methyl benzoate, 2,4-dihydroxy,methyl b-resorcylate
16523
methyl 2,4-dihydroxybenzoate
COC(=O)C1=C(C=C(C=C1)O)O
2150-47-2
C8H8O4
168.148
MFCD00002276
IIFCLXHRIYTHPV-UHFFFAOYSA-N
2,4-dihydroxybenzoic acid methyl ester,benzoic acid, 2,4-dihydroxy-, methyl ester,methyl beta-resorcylate,methyl-2,4-dihydroxybenzoate,unii-xe68bu3oyj,xe68bu3oyj,.beta.-resorcylic acid, methyl ester,beta-resorcylic acid methyl ester,methyl benzoate, 2,4-dihydroxy,methyl b-resorcylate
16523
methyl 2,4-dihydroxybenzoate
COC(=O)C1=C(C=C(C=C1)O)O
Applications
Methyl 2,4-dihydroxybenzoate is used as a cosmetic agents. And it is used in synthesis of raw material for fine chemicals, organic intermediate, pharmaceutical intermediate.

Solubility
Soluble in methanol.

Notes
Keep container tightly closed. Store away from oxidizing agents.
Quantity 100g
Melting Point 116°C to 119°C
Formula Weight 168.15
Percent Purity 97%
Chemical Name or Material Methyl 2,4-dihydroxybenzoate

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

TSCA : TSCA